AXYFX250LR
MAXYMum Recovery® pipette tips for Beckman® FX Robotics System
capacity 250 μL, pkg of 5x960tips/cs, clear
Synonym(s):
pipet tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
volume:
250 μL
packaging:
pkg of 5x960tips/cs
manufacturer/tradename:
Axygen Inc FX-250-L-R
Corning
Corning
manufacturer/tradename
Axygen Inc FX-250-L-R
Corning
packaging
pkg of 5x960tips/cs
capacity
250 μL
volume
250 μL
color
clear
Looking for similar products? Visit Product Comparison Guide
Other Notes
Browse all pipette tips
Legal Information
Beckman is a registered trademark of Beckman Coulter, Inc.
Maxymum Recovery is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service