Skip to Content
Merck
All Photos(3)

Key Documents

AV40528

Sigma-Aldrich

Anti-SFRS10 (AB1) antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

mol wt

32 kDa

species reactivity

canine, rat, human, rabbit, horse, mouse, pig, bovine

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

storage temp.

−20°C

Gene Information

human ... SFRS10(6434)

General description

Transformer-2 protein homolog β/splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila) (TRA2B, SFRS10) is an alternative splicing regulator that itself exists as alternative splice variants. Alternative mRNA splicing provides transcript diversity. SFRS10 binds SMN1/SMN2 RNA and restores full-length (FL)-SMN2 mRNA levels in vitro. Deficiency of SFRS10 results in early embryonic lethality.

Specificity

Anti-SFRS10 (AB1) polyclonal antibody reacts with canine, chicken, human, mouse, rat, pig, and bovine transformer-2 protein homolog β/splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila) proteins.

Immunogen

The immunogen for anti-SFRS10 antibody: synthetic peptide derected towards the N terminal of human SFRS10

Application

Anti-SFRS10 (AB1) polyclonal antibody is used to tag transformer-2 protein homolog β/splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila) for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of transformer-2 protein homolog β/splicing factor, arginine/serine-rich 10 (transformer 2 homolog, Drosophila) in the regulation of alternative pre-mRNA splicing.

Sequence

Synthetic peptide located within the following region: MSDSGEQNYGERESRSASRSGSAHGSGKSARHTPARSRSKEDSRRSRSKS

Physical form

Lyophilized from PBS buffer with 2% sucrose

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yupu Diao et al.
Cerebral cortex (New York, N.Y. : 1991), 28(4), 1168-1182 (2017-03-24)
The dorsal Lateral Geniculate Nucleus (dLGN) is the primary image-forming target of the retina and shares a reciprocal connection with primary visual cortex (V1). Previous studies showed that corticothalamic input is essential for the development of thalamocortical projections, but less
Somsakul Pop Wongpalee et al.
eLife, 5 (2016-11-25)
Polypyrimidine-tract binding protein PTBP1 can repress splicing during the exon definition phase of spliceosome assembly, but the assembly steps leading to an exon definition complex (EDC) and how PTBP1 might modulate them are not clear. We found that PTBP1 binding
Devi Krishna Priya Karunakaran et al.
Gene expression patterns : GEP, 13(8), 425-436 (2013-08-13)
Processing of mRNAs including, alternative splicing (AS), mRNA transport and translation regulation are crucial to eukaryotic gene expression. For example, >90% of the genes in the human genome are known to undergo alternative splicing thereby expanding the proteome production capacity
Thomas C Whisenant et al.
PloS one, 10(12), e0144409-e0144409 (2015-12-08)
Activation of CD4 T cells is a reaction to challenges such as microbial pathogens, cancer and toxins that defines adaptive immune responses. The roles of T cell receptor crosslinking, intracellular signaling, and transcription factor activation are well described, but the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service