Skip to Content
Merck
All Photos(4)

Key Documents

AV39521

Sigma-Aldrich

Anti-TEAD1 (AB1) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-TEA domain family member 1 (SV40 transcriptional enhancer factor)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€498.00

€498.00


Please contact Customer Service for Availability


Select a Size

Change View
100 μL
€498.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€498.00


Please contact Customer Service for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

46 kDa

species reactivity

horse, bovine, rabbit, human, rat, guinea pig, mouse, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TEAD1(7003)

General description

TEAD1 is a transcriptional factor that blocks the expression of prolactin in human uterine decidual cells. TEAD1 mutation has been linked to Sveinsson′s chorioretinal atrophy.
Rabbit Anti-TEAD1 antibody recognizes pig, zebrafish, human, mouse, rat, bovine, and canine TEAD1.

Immunogen

Synthetic peptide directed towards the C terminal region of human TEAD1

Application

Rabbit Anti-TEAD1 antibody is suitable for western blot applications at a concentration of 0.25 μg/ml and for IHC at 16 μg/ml.

Biochem/physiol Actions

TEAD1 is a transcriptional enhancer. It interacts with a muscle-specific cofactor to promote skeletal muscle gene expression. The mutation in the TEAD1 gene is the cause of Sveinsson′s chorioretinal atrophy

Sequence

Synthetic peptide located within the following region: YMMNSVLENFTILLVVTNRDTQETLLCMACVFEVSNSEHGAQHHIYRLVK

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ragnheidur Fossdal et al.
Human molecular genetics, 13(9), 975-981 (2004-03-16)
Sveinsson's chorioretinal atrophy (SCRA), also referred to as helicoid peripapillary chorioretinal degeneration or atrophia areata, is an autosomal dominant eye disease, characterized by symmetrical lesions radiating from the optic disc involving the retina and the choroid. Genome-wide linkage analysis mapped
Cherie A Kessler et al.
Molecular and cellular endocrinology, 295(1-2), 32-38 (2008-09-09)
Forced overexpression of TEAD1 in human uterine fibroblast (HUF) and human endometrial stromal cells markedly inhibited prolactin promoter activity in both cell types in a dose-dependent manner, with maximal inhibition of greater than 90%. Conversely, the knockdown of TEAD1 expression
Shohei Ikeda et al.
JACC. Basic to translational science, 4(5), 611-622 (2019-11-27)
Patients with diabetes are more prone to developing heart failure in the presence of high blood pressure than those without diabetes. Yes-associated protein (YAP), a key effector of the Hippo signaling pathway, is persistently activated in diabetic hearts, and YAP

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service