Skip to Content
Merck
All Photos(1)

Key Documents

A8754

Sigma-Aldrich

Methyl α-aminoisobutyrate hydrochloride

Synonym(s):

α-Aminoisobutyric acid methyl ester hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11NO2 · HCl
CAS Number:
Molecular Weight:
153.61
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (TLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white

storage temp.

−20°C

SMILES string

Cl.COC(=O)C(C)(C)N

InChI

1S/C5H11NO2.ClH/c1-5(2,6)4(7)8-3;/h6H2,1-3H3;1H

InChI key

NVWZNEDLYYLQJC-UHFFFAOYSA-N

Biochem/physiol Actions

Methyl α-aminoisobutyrate is used as a specific substrate of amino acid transport system A in studies on amino acid transport systems.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oak Z Chi et al.
Neurochemical research, 34(7), 1249-1254 (2009-01-08)
This study was performed to determine whether exogenous N-methyl-D: -aspartate (NMDA) or alpha-amino-3-hydroxy-5-methyl-4-isoxazole propionic acid (AMPA) would aggravate blood-brain barrier (BBB) disruption in focal cerebral ischemia in rats. Forty-five minutes after middle cerebral artery (MCA) occlusion, one of the following
A Gadea et al.
Glia, 26(4), 273-279 (1999-06-26)
Rapid termination of the synaptic action of glutamate (Glu) and glycine (Gly) is achieved by uptake into the presynaptic terminal and glial cells. In the vertebrate CNS, Gly acts both as an inhibitory neurotransmitter and as a Glu modulator or
J W Tsang et al.
Journal of medicinal chemistry, 27(12), 1663-1668 (1984-12-01)
Stereochemical and structural aspects of the variations in the C-terminal residue of L-aspartyl-L-phenylalanine methyl ester have been investigated. Novel configurational analogues such as L-aspartyl-D-alanine benzyl ester and L-aspartyl-D-alpha-aminobutyric acid benzyl ester were found to be sweet. In addition, chiral and
Koichi Kato et al.
Bioorganic & medicinal chemistry letters, 21(8), 2437-2440 (2011-03-12)
We describe the synthesis of (11)C-labeled α-aminoisobutyric acid 2 from iodo[(11)C]methane and methyl N-(diphenylmethylen)-d,l-alaniate (5). The tetrabutylammonium fluoride (TBAF)-promoted α-[(11)C]methylation of sterically hindered analog 5 was a key step in our synthesis process. Total radiochemical conversion of 2 was high

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service