Skip to Content
Merck
All Photos(1)

Key Documents

A6351

Sigma-Aldrich

A-315456

>98% (HPLC), solid

Synonym(s):

N-[3-(cyclohexylidene(1H-imidazol-4-ylmethyl)phenyl]ethanesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H23N3O2S
Molecular Weight:
345.46
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

>98% (HPLC)

form

solid

color

off-white

solubility

DMSO: soluble 24 mg/mL
H2O: insoluble

SMILES string

CCS(=O)(=O)Nc1cccc(c1)C(=C2/CCCCC2)\c3cnc[nH]3

InChI

1S/C18H23N3O2S/c1-2-24(22,23)21-16-10-6-9-15(11-16)18(17-12-19-13-20-17)14-7-4-3-5-8-14/h6,9-13,21H,2-5,7-8H2,1H3,(H,19,20)

InChI key

MTZVJHSZYMWIAG-UHFFFAOYSA-N

Gene Information

human ... ADRA1D(146)

Application

A-315456 is a potent and selective α1D-antagonist that may serve as a useful pharmacological ligand to probe the physiological role of the α(1D)-adrenoceptor subtype in normal and disease states.

Biochem/physiol Actions

Selective α1D-adrenoceptor antagonist.

Features and Benefits

This compound was developed by Abbott. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S A Buckner et al.
European journal of pharmacology, 433(1), 123-127 (2002-01-05)
In functional assays, A-315456, N-[3-(cyclohexylidene-(1H-imidazol-4-ylmethyl))phenyl]ethanesulfonamide, behaved as an alpha(1D)-adrenoceptor subtype selective antagonist (pA(2)=8.34) in the rat aorta. It was 83-fold less potent at the alpha(1B)-adrenoceptor subtype expressed in the rat spleen, and was inactive at the alpha(1A)-adrenoceptor subtype expressed in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service