Skip to Content
Merck
All Photos(1)

Key Documents

A2269

Sigma-Aldrich

2-Acetamidofluorene

Synonym(s):

N-(2-Fluorenyl)acetamide, 2-AAF, 2-Acetamidofluorene, N-Acetyl-2-aminofluorene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13NO
CAS Number:
Molecular Weight:
223.27
Beilstein:
2807677
EC Number:
MDL number:
UNSPSC Code:
12352200

mp

192-196 °C (lit.)

SMILES string

CC(=O)Nc1ccc-2c(Cc3ccccc-23)c1

InChI

1S/C15H13NO/c1-10(17)16-13-6-7-15-12(9-13)8-11-4-2-3-5-14(11)15/h2-7,9H,8H2,1H3,(H,16,17)

InChI key

CZIHNRWJTSTCEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

A genotoxic carcinogen that is used to model liver carcinogenesis in rat. When N-hydroxylated by cytochrome CYP1A2 in the liver, 2-AAF forms adducts with DNA and is tumorigenic in liver and bladder.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshifumi Uno et al.
Mutation research. Genetic toxicology and environmental mutagenesis, 786-788, 182-187 (2015-07-28)
The data from the JaCVAM-organized international validation study of the in vivo rat alkaline comet assay were reported and analyzed statistically using the simple means of % tail DNA. However, OECD test guideline TG 489 recommends use of the median
Yoshifumi Uno et al.
Mutation research. Genetic toxicology and environmental mutagenesis, 786-788, 45-76 (2015-07-28)
The in vivo rodent alkaline comet assay (comet assay) is used internationally to investigate the in vivo genotoxic potential of test chemicals. This assay, however, has not previously been formally validated. The Japanese Center for the Validation of Alternative Methods
A R Kraynak et al.
Mutation research. Genetic toxicology and environmental mutagenesis, 786-788, 77-86 (2015-07-28)
As part of the Japanese Center for the Validation of Alternative Methods (JaCVAM) initiative international validation study of the in vivo rat alkaline comet assay (comet assay), we examined the ability of the assay to determine the genotoxicity of 2-acetylaminofluorene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service