Skip to Content
Merck
All Photos(1)

Key Documents

A1340

Millipore

Antibiotic assay medium No. 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

pH

7.0±0.2 (25 °C)

Application

Used in the performance of antibiotic assays.

Other Notes

Components in g/L:
Peptic Digest of Animal Tissue, 5.00
Yeast Extract, 1.50
Beef Extract, 1.50
Dextrose, 1.00
Sodium Chloride, 3.50
Dipotassium Phosphate, 3.68
Potassium Dihydrogen Phosphate, 1.32

replaced by

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tests and Methods of Assay of Antibiotics and Antibiotic Containing Drugs, 21, 436-436 (1983)
USP 23/NF18.
United States Pharmacopeia/National Formulary (1995)
USP 22/NF 17.
United States Pharmacopeia/National Formulary (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service