99072804
T-3-CI-2-0.fl cell line
mouse spleen, Lymphoblastoid-like
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
NACRES:
NA.81
Recommended Products
Product Name
T-3-CI-2-0.fl cell line,
biological source
mouse spleen
Quality Level
form
liquid
growth mode
Suspension
karyotype
Not specified
morphology
Lymphoblastoid-like
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse friend leukemia lymphoblast
Cell Line Description
T-3-Cl-2-0.fl is a clone of T-3-Cl-2, which originated from a splenic lesion of Friend virus-induced leukemia in a DD mouse. The cell line can be induced to differentiate into erythroblastoid cells with 1.5% DMSO. T-3-Cl-2-0.fl has been eradicated from mycoplasma using MC-210 by the originator.
Application
Study of erythroid differentiation
Culture Medium
RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Maintain cultures between 3-9 x 100,000 cells/ml; 5%CO2; 37ºC
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service