Skip to Content
Merck
All Photos(1)

Key Documents

77630

Sigma-Aldrich

Phenolphthalein bisphosphate pyridine salt

≥90% (T)

Synonym(s):

Phenolphthalein diphosphate pyridine salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16O10P2 · C5H5N
CAS Number:
Molecular Weight:
557.38
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Assay

≥90% (T)

form

powder

impurities

≤0.05% free phenolphthalein

loss

≤2% loss on drying

storage temp.

2-8°C

SMILES string

c1ccncc1.OP(O)(=O)Oc2ccc(cc2)C3(OC(=O)c4ccccc34)c5ccc(OP(O)(O)=O)cc5

InChI

1S/C20H16O10P2.C5H5N/c21-19-17-3-1-2-4-18(17)20(28-19,13-5-9-15(10-6-13)29-31(22,23)24)14-7-11-16(12-8-14)30-32(25,26)27;1-2-4-6-5-3-1/h1-12H,(H2,22,23,24)(H2,25,26,27);1-5H

InChI key

DBXHFBVMRQOQKW-UHFFFAOYSA-N

Application

Suitable as substrate for phosphatase.

Other Notes

Chromogenic substrate for acid and alkaline phosphatases[1][2][3][4][5]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Changes in the phosphatase activity of Baker's yeast during the growth phase and location of the phosphatases in the yeast cell.
H SUOMALAINEN et al.
Biochimica et biophysica acta, 37, 482-490 (1960-01-29)
One-step automated alkaline phosphatase analysis.
D Comfort et al.
Clinica chimica acta; international journal of clinical chemistry, 14(2), 263-269 (1966-08-01)
J J Thrasher et al.
Journal - Association of Official Analytical Chemists, 64(1), 196-198 (1981-01-01)
A chemical method was developed to determine the presence of mammalian feces in food. The method consisted of the colorimetric determination of alkaline phosphatase and involved the splitting of the phosphate radical and phenolphthalein diphosphate and the resultant color development
A simple and reliable method for estimation of alkaline phosphatase in tissue homogenates.
J P Manning et al.
Enzymologia, 31(5), 309-320 (1966-11-30)
Microdetermination of phosphatases employing phenolphthalein diphosphate as substrate.
J Fischl et al.
Clinical chemistry, 13(11), 941-946 (1967-11-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service