Skip to Content
Merck
All Photos(1)

Key Documents

72827

Sigma-Aldrich

Mobile Phase A for separation of amino acids on ABI® sequencers

Synonym(s):

Tetrahydrofuran solution

Sign Into View Organizational & Contract Pricing


About This Item

Beilstein:
3668542
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

suitability

in accordance for sequence analysis test

storage temp.

2-8°C

SMILES string

C1CCOC1

Looking for similar products? Visit Product Comparison Guide

Application

1 liter needs to be supplemented by 8-10 mL Premix (79923). In case of coelution of aspartic acid with DTT artifact peak, 100 μl TFA (09653) should be added.

Legal Information

ABI PRISM is a registered trademark of Applera Corporation or its subsidiaries in the US and/or certain other countries

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilaria Massarelli et al.
European journal of medicinal chemistry, 44(9), 3658-3664 (2009-03-11)
A dataset comprising 55 chemicals with hepatocarcinogenic potency indices was collected from the Carcinogenic Potency Database with the aim of developing QSAR models enabling prediction of the above unwanted property for New Chemical Entities. The dataset was rationally split into
Anita Toulmin et al.
Journal of medicinal chemistry, 51(13), 3720-3730 (2008-06-19)
Partition coefficients were measured for 47 compounds in the hexadecane/water ( P hxd) and 1-octanol/water ( P oct) systems. Some types of hydrogen bond acceptor presented by these compounds to the partitioning systems are not well represented in the literature
Alfonso Pérez-Garrido et al.
Bioorganic & medicinal chemistry, 17(2), 896-904 (2008-12-06)
This paper reports a QSAR study for predicting the complexation of a large and heterogeneous variety of substances (233 organic compounds) with beta-cyclodextrins (beta-CDs). Several different theoretical molecular descriptors, calculated solely from the molecular structure of the compounds under investigation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service