Skip to Content
Merck
All Photos(1)

Documents

63189

Sigma-Aldrich

Maleic acid

tested according to Ph. Eur.

Synonym(s):

Acidum maleicum, cis-Butenedioic acid, Toxilic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO2CCH=CHCO2H
CAS Number:
Molecular Weight:
116.07
Beilstein:
605762
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Agency

USP/NF
tested according to Ph. Eur.

Assay

99.0-101.0% substance (anhydrous) basis

form

fine crystals
powder

mp

130-135 °C (lit.)

solubility

water: soluble 478.8 g/L at 20 °C

density

1.59 g/mL at 25 °C (lit.)

cation traces

Fe: ≤5 ppm
heavy metals: ≤10 ppm

application(s)

pharmaceutical (small molecule)

storage temp.

2-8°C

SMILES string

OC(=O)\C=C/C(O)=O

InChI

1S/C4H4O4/c5-3(6)1-2-4(7)8/h1-2H,(H,5,6)(H,7,8)/b2-1-

InChI key

VZCYOOQTPOCHFL-UPHRSURJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Maleic acid is a dicarboxylic acid. It can be synthesized from crotonaldehyde.

Application

Maleic acid may be used in the synthesis of (R,S)-malic acid and fumaric acid. It may be employed as an anionic monomer in the preparation of superswelling acrylamide (AAm)/maleic acid (MA) hydrogels.†

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eagleson M.
Concise Encyclopedia Chemistry, 277-277 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 617-617 (1994)
Eagleson M.
Concise Encyclopedia Chemistry, 434-434 (1994)
J M Herreras et al.
Ophthalmology, 99(7), 1082-1088 (1992-07-01)
This study was undertaken to see whether long-term locally applied ocular medications produced any alterations in the ocular surface, and, in particular, whether it caused damage to the mucus layer of the tear film. The authors studied the ocular surface
Hanna Kotikoski et al.
Acta ophthalmologica Scandinavica, 81(4), 355-360 (2003-07-16)
To investigate whether a nitric oxide donor given as a single oral dose is able to modify aqueous humour flow in healthy volunteers. Ten healthy volunteers participated in a randomized, double-masked and placebo-controlled cross-over study. Aqueous humour flow was assessed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service