Skip to Content
Merck
All Photos(1)

Key Documents

61967

Sigma-Aldrich

Chromeo 642-Azide

for click labeling

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

form

solid

solubility

DMF: 0.5 mg/mL, clear, blue

fluorescence

λex 646 nm±5 nm; λem 666 nm±5 nm in PBS, pH 7.4

storage temp.

−20°C

General description

C38H52N6O4P(Br) MW 687 (767 including Br- anion)

Application

Reactive fluorescent Chromeo Dyes serve as bright labels for antibodies and other biomolecules, enabling detection in immunofluorescence, high content screening, ELISA, FRET applications or flow cytometry. Chromeo 642 may be used in place of Cy5 dye. Chromeo 642-Azide is provided for click labeling applications.

Legal Information

Chromeo is a trademark of Active Motif Chromeon GmbH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lei Chen et al.
The Journal of organic chemistry, 73(21), 8279-8285 (2008-10-02)
In search of an appropriate position for the fluorescent labeling, six chemically available positions of the flavonone core of naringenin have been examined. A number of azido-containing naringenin derivatives were accordingly prepared in various site-specific fashions, and the mild Cu(I)-catalyzed
Ziyang Hao et al.
Chemical communications (Cambridge, England), 47(15), 4502-4504 (2011-03-10)
A concise route was developed for the facile synthesis of a cyclic pyrrolysine analogue bearing an azide handle. Directed evolution enabled the encoding of this non-natural amino acid in both prokaryotic and eukaryotic cells, which offers a highly efficient approach
Yuri Motorin et al.
Nucleic acids research, 39(5), 1943-1952 (2010-11-03)
This work identifies the combination of enzymatic transfer and click labeling as an efficient method for the site-specific tagging of RNA molecules for biophysical studies. A double-activated analog of the ubiquitous co-substrate S-adenosyl-l-methionine was employed to enzymatically transfer a five
Migle Tomkuviene et al.
Nucleic acids research, 40(14), 6765-6773 (2012-05-09)
Biophysical and mechanistic investigation of RNA function requires site-specific incorporation of spectroscopic and chemical probes, which is difficult to achieve using current technologies. We have in vitro reconstituted a functional box C/D small ribonucleoprotein RNA 2'-O-methyltransferase (C/D RNP) from the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service