Skip to Content
Merck
All Photos(1)

Key Documents

3533-010-02

Sigma-Aldrich

Cultrex® Reduced Growth Factor BME, Type 2 PathClear® (2 x 5ml)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
Pricing and availability is not currently available.

concentration

12-18 mg/mL

shipped in

dry ice

storage temp.

−70°C

General description

Basement membranes are continuous sheets of specialized extracellular matrix that form an interface between endothelial, epithelial, muscle, or neuronal cells and their adjacent stroma. Basement membranes are degraded and regenerated during development and wound healing. They not only support cells and cell layers, but they also play an essential role in tissue organization that affects cell adhesion, migration, proliferation, and differentiation. Basement membranes provide major barriers to invasion by metastatic tumor cells. Cultrex® Basement Membrane Extract (BME) is a soluble form of basement membrane purified from Engelbreth-Holm-Swarm (EHS) tumor. The extract gels at 37°C to form a reconstituted basement membrane. The major components of BME include laminin, collagen IV, entactin, and heparin sulfate proteoglycan. BME can be used in a multiple applications, under a variety of cell culture conditions, for maintaining growth or promoting differentiation of primary endothelial, epithelial, smooth muscle and stem cells. BME can also be utilized in cell attachment, neurite outgrowth, angiogenesis, in vitro cell invasion and in vivo tumorigenicity assays. Recently we have developed two additional formulations of Cultrex® BME known as Cultrex® BME Type 2 and Cultrex® BME Type 3. Cultrex® BME Type 2 provides a proprietary formulation that is higher in tensile strength when compared to our original BME, while Cultrex® BME Type 3 is physiologically aligned with the in vivo solid tumors environment and is recommended for xenografts and other in vivo applications.

Physical form

Solution in Dulbecco′s Modified Eagle′s medium without phenol red, with 10 μg/ml gentamicin sulfate.

Other Notes

Sterility Testing:
PathClear® - Negative by PCR test for mycoplasma; 17 bacterial and virus strains typically included in mouse antibody production (MAP) testing, plus 13 additional murine infectious agents including LDEV, for a total of 31 organisms and viruses.No bacterial or fungal growth detected after incubation at 37°C for 14 days following USP sterility testing guidelines. Endotoxin concentration = 8 EU/ml by LAL assay.
Storage Conditions and Product Stability:
Product is stable for a minimum of 3 months from date of shipment when stored at –20°C in a manual defrost freezer. For optimal stability, store at –80°C. Avoid freeze-thaw cycles.

Legal Information

Cultrex is a registered trademark of Trevigen, Inc.
PathClear is a registered trademark of Trevigen, Inc.

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Norman Sachs et al.
Cell, 172(1-2), 373-386 (2017-12-12)
Breast cancer (BC) comprises multiple distinct subtypes that differ genetically, pathologically, and clinically. Here, we describe a robust protocol for long-term culturing of human mammary epithelial organoids. Using this protocol, >100 primary and metastatic BC organoid lines were generated, broadly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service