Key Documents
2501S
Millipore
BIOEAZE EZ Top and Aseptic Connecting Devices
About This Item
UNSPSC Code:
12352200
description
Designed to Aseptically Transfer of Liquids into or from Bottles by Quick Connects
sterility
sterile; γ-irradiated
General description
Fits 500 mL or 1 L Bottle with 082 Clear C-Flex® Stopper with 38 mm Cap with 1 x 2 ft. length of 3/16" x 3/8" Clear 082 C-Flex® Tubing with Pinch Clamp and 1/4" Female Polycarbonate Quick Connect with Cap
Packaging
Double Polybagged
Legal Information
BIOEAZE is a trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)