Skip to Content
Merck
All Photos(1)

Key Documents

05803

Sigma-Aldrich

N-(2-Aminoethyl)rhodamine 6G-amide bis(trifluoroacetate)

≥95.0% (HPCE)

Synonym(s):

Rhodamine 6G-N-(2-aminoethyl)amide bis(trifluoroacetate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H32N4O2·C4H2F6O4
CAS Number:
Molecular Weight:
684.63
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Assay

≥95.0% (HPCE)

form

powder

fluorescence

λex 528 nm; λem 553 nm in 0.1 M phosphate pH 3.0

storage temp.

2-8°C

SMILES string

OC(=O)C(F)(F)F.OC(=O)C(F)(F)F.CCNc1cc2OC3=CC(=N\CC)\C(C)=CC3=C(c2cc1C)c4ccccc4C(=O)NCCN

InChI

1S/C28H32N4O2.2C2HF3O2/c1-5-30-23-15-25-21(13-17(23)3)27(19-9-7-8-10-20(19)28(33)32-12-11-29)22-14-18(4)24(31-6-2)16-26(22)34-25;2*3-2(4,5)1(6)7/h7-10,13-16,30H,5-6,11-12,29H2,1-4H3,(H,32,33);2*(H,6,7)/b31-24-;;

InChI key

GBBDSCJKOGANEP-MDRFVTBBSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service