Skip to Content
Merck
All Photos(1)

Key Documents

02187

Sigma-Aldrich

Isoorientin

≥98.0% (HPLC)

Synonym(s):

Homoorientin, Luteolin 6-C-β-D-glucoside, Luteolin 6-C-glucoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20O11
CAS Number:
Molecular Weight:
448.38
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

Assay

≥98.0% (HPLC)

SMILES string

OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)c2c(O)cc3OC(=CC(=O)c3c2O)c4ccc(O)c(O)c4

InChI

1S/C21H20O11/c22-6-14-17(27)19(29)20(30)21(32-14)16-11(26)5-13-15(18(16)28)10(25)4-12(31-13)7-1-2-8(23)9(24)3-7/h1-5,14,17,19-24,26-30H,6H2/t14-,17-,19+,20-,21+/m1/s1

InChI key

ODBRNZZJSYPIDI-VJXVFPJBSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

C-glycosyl flavone with anti-inflammatory, antimicrobial and antioxidant properties. Induces antioxidant response through PI3K singaling.
Luteolin, a common flavonoid, has been shown to inhibit secretion of ET-1 by porcine aortic endothelial cells at a 10 uM concentration. However, the luteolin glycoside compound, luteolin 6-C-glucoside, showed very weak inhibition of ET-1 release and inhibition only occurred at higher concentrations of luteolin 6-C-glucoside.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luteolin inhibits endothelin-1 secretion in cultured endothelial cells.
Kozakai, T.
Biosciences, Biotechnology Research Asia, 69, 1613-1615 (2005)
Jinyong Peng et al.
Journal of chromatography. A, 1074(1-2), 111-115 (2005-06-09)
High-speed counter-current chromatography (HSCCC) with a solvent system composed of ethyl acetate-n-butanol-water (2:1:3, v/v/v) was used to isolate and separate two C-glycosylflavones from Patrinia villosa Juss, a traditional Chinese medicine. The separation produced 42.9 mg isovitexin and 20.1 mg isoorientin
Dong-Seon Kim et al.
Planta medica, 76(7), 678-682 (2009-12-05)
We aimed to find antiallergic agents from natural sources using mast cells activated during allergic reaction. We screened over 2000 plants for blockade of histamine release and identified two of them, S. baicalenesis and P. edulis. Bioassay-guided fractionation led to
Mala Ranghoo-Sanmukhiya et al.
Phytochemical analysis : PCA, 21(6), 566-574 (2010-09-08)
Aloe tormentorii, A. purpurea and A. macra are used as multipurpose folk medicines in Réunion and Mauritius Islands and are mistaken for the introduced Aloe vera. To compare the phytochemical, antimicrobial and DNA profiles of Aloe endemic to Mauritius and
Severina Pacifico et al.
Journal of natural products, 73(12), 1973-1978 (2010-11-18)
Eight flavonoid C-glycosides, including three new analogues, have been isolated from leaf and root methanolic extracts of Petrorhagia velutina, a Mediterranean herbaceous plant. The antiproliferative activity against human hepatoblastoma cancer cell line HepG2 has been analyzed by the MTT (3-(4,5-dimethyl-2-thiazolyl)-2,5-diphenyl-2H-tetrazolium

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service