Skip to Content
Merck
All Photos(1)

Key Documents

Y0001891

Sildenafil for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Sildenafil citrate salt, 1-[[3-(4,7-Dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-4-ethoxyphenyl]sulfonyl]-4-methylpiperazine citrate salt, 5-[2-Ethoxy-5-(4-methylpiperazin-1-yl)sulfonylphenyl]-1-methyl-3-propyl-4H-pyrazolo[5,4-e]pyrimidin-7-one citrate salt, Revatio, UK-92,480

Sign Into View Organizational & Contract Pricing

Select a Size

15 MG
€181.00

€181.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
15 MG
€181.00

About This Item

Empirical Formula (Hill Notation):
C22H30N6O4S ·C6H8O7
CAS Number:
Molecular Weight:
666.70
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€181.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

sildenafil

manufacturer/tradename

EDQM

format

neat

SMILES string

OC(=O)CC(O)(CC(O)=O)C(O)=O.CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)N4CCN(C)CC4

InChI

1S/C22H30N6O4S.C6H8O7/c1-5-7-17-19-20(27(4)25-17)22(29)24-21(23-19)16-14-15(8-9-18(16)32-6-2)33(30,31)28-12-10-26(3)11-13-28;7-3(8)1-6(13,5(11)12)2-4(9)10/h8-9,14H,5-7,10-13H2,1-4H3,(H,23,24,29);13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)

InChI key

DEIYFTQMQPDXOT-UHFFFAOYSA-N

Gene Information

human ... PDE5A(8654)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including MSDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Sildenafil is a potent, selective inhibitor of cGMP specific phosphodiesterase type 5 (PDE5). Sildenafil is used to treat erectile dysfunction and pulmonary arterial hypertension. NO activates guanylate cyclase, which results in increased levels of cGMP, producing smooth muscle relaxation. Sildenafil enhances the effect of NO by inhibiting PDE5, which is responsible for degradation of cGMP.
Sildenafil is a potent, selective inhibitor of cGMP specific phosphodiesterase type 5 (PDE5). Sildenafil is used to treat erectile dysfunction and pulmonary hypertension.

Packaging

Unit quantity: 15 mg. Subject to change. The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service