Skip to Content
Merck
All Photos(1)

Documents

W509348

Sigma-Aldrich

Diethyl adipate

≥99%, FG

Synonym(s):

Hexanedioic acid 1,6-diethyl ester, NSC 19160, NSC 3363

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCO(CH2)4COOC2H5
CAS Number:
Molecular Weight:
202.25
Beilstein:
1780035
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
09.348

biological source

synthetic

Quality Level

grade

FG
Fragrance grade
Kosher

Agency

follows IFRA guidelines

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 872/2012

Assay

≥99%

refractive index

n20/D 1.427 (lit.)

bp

251 °C (lit.)

mp

−20-−19 °C (lit.)

density

1.009 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

SMILES string

CCOC(=O)CCCCC(=O)OCC

InChI

1S/C10H18O4/c1-3-13-9(11)7-5-6-8-10(12)14-4-2/h3-8H2,1-2H3

InChI key

VIZORQUEIQEFRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service