Skip to Content
Merck
All Photos(3)

Key Documents

VT000102

Millipore

Enterococcus faecium WDCM 00010 Vitroids

15-80 CFU mean value range, certified reference material, suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

grade

certified reference material

form

disc (hydrosoluble)

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

microbiology

storage temp.

−20°C

suitability

Enterococcus spp.

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
Download your certificate at: http://www.sigma-aldrich.com.

Please specify if you require a single batch - in the remark field when ordering (only for orders with more than 15 packs)

Other Notes

ATCC® 19434, NCTC 7171, NCPF -, DSM 20477, LMG 11423, IHEM -, CIP 103014T
Origin Strain No. CECT® 410

Legal Information

Derived from CECT® strains
ATCC is a registered trademark of American Type Culture Collection
CECT is a registered trademark of Universitat de Valencia
Vitroids is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service