Skip to Content
Merck
All Photos(1)

Key Documents

T6159

Supelco

Trimebutine

analytical standard

Synonym(s):

3,4,5-Trimethoxybenzoic acid 2-(dimethylamino)-2-phenylbutyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H29NO5
CAS Number:
Molecular Weight:
387.47
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

CCC(COC(=O)c1cc(OC)c(OC)c(OC)c1)(N(C)C)c2ccccc2

InChI

1S/C22H29NO5/c1-7-22(23(2)3,17-11-9-8-10-12-17)15-28-21(24)16-13-18(25-4)20(27-6)19(14-16)26-5/h8-14H,7,15H2,1-6H3

InChI key

LORDFXWUHHSAQU-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kwan Hyung Cho et al.
International journal of pharmaceutics, 400(1-2), 145-152 (2010-09-10)
To develop a novel combination tablet which contained 100 mg trimebutine maleate and 5 mg mosapride citrate (TMCT) for the treatment of functional dyspepsia, the wet granulation method was used to prepare TMCTs with various amounts of diluents and stabilizers.
O Bekem et al.
Acta gastro-enterologica Belgica, 68(4), 416-418 (2006-01-26)
Of the feeding disorders in children, poor appetite is probably one of the most common complaints notified by parents. Since gastric motility disorders may be the cause of this symptom, the aim of our study was to investigate the effect
A V Krymchantowski et al.
Cephalalgia : an international journal of headache, 26(7), 871-874 (2006-06-17)
Gastroparesis frequently happens during migraine attacks, postponing the onset of action of orally administered drugs. Furthermore, triptans seem to work better in the earlier phases of the migraine attacks. Therefore, associating a gastrokinetic drug with a triptan may translate into
Imke Masuy et al.
Alimentary pharmacology & therapeutics, 49(9), 1134-1172 (2019-03-30)
Functional dyspepsia, consisting of epigastric pain syndrome and postprandial distress syndrome, is a prevalent functional gastrointestinal disorder. To date, only limited treatment options are available and conflicting results in terms of efficacy have been reported. Consequently, nonpharmacological treatment options are
[Trimebutine. Pharmacology, recent concepts about its mechanism of action, therapeutic results].
B Fraitag et al.
Gastroenterologie clinique et biologique, 16(5), 413-420 (1992-01-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service