Skip to Content
Merck
All Photos(1)

Documents

STS0030

Sigma-Aldrich

Digitonin

BioUltra, used as non-ionic detergent

Synonym(s):

Digitin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C56H92O29
CAS Number:
Molecular Weight:
1229.31
Beilstein:
78654
MDL number:
UNSPSC Code:
12161900
NACRES:
NA.21

Quality Level

product line

BioUltra

mol wt

micellar avg mol wt 70,000

aggregation number

60

CMC

<0.5 mM (20-25°C)

mp

230-240 °C (dec.) (lit.)

SMILES string

C[C@@]12[C@]([C@@H]3C)([H])[C@](O[C@]34CC[C@@H](C)CO4)([H])[C@@H](O)[C@@]1([H])[C@@](CC[C@]5([H])[C@@]6(C[C@@H](O)[C@H](O[C@]([C@@H]([C@@H](O)[C@H]7O[C@@](O[C@H](CO)[C@@H](O)[C@@H]8O[C@@](OC[C@@H](O)[C@@H]9O)([H])[C@@H]9O)([H])[C@@H]8O[C@@](O[C@H](CO)[C@H

InChI

1S/C56H92O29/c1-19-7-10-56(75-17-19)20(2)31-45(85-56)37(67)32-22-6-5-21-11-26(24(61)12-55(21,4)23(22)8-9-54(31,32)3)76-50-42(72)39(69)44(30(16-60)80-50)81-53-48(47(36(66)29(15-59)79-53)83-49-40(70)33(63)25(62)18-74-49)84-52-43(73)46(35(65)28(14-58)78-52)82-51-41(71)38(68)34(64)27(13-57)77-51/h19-53,57-73H,5-18H2,1-4H3/t19-,20+,21+,22-,23+,24-,25-,26-,27-,28-,29-,30-,31+,32-,33+,34-,35+,36-,37+,38+,39-,40-,41-,42-,43-,44+,45-,46+,47+,48-,49+,50-,51+,52+,53+,54-,55+,56-/m1/s1

InChI key

UVYVLBIGDKGWPX-KUAJCENISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Digitonin, a steroidal saponin extracted from the purple foxglove plant (Digitalis purpurea), stands as a versatile non-ionic detergent crucial for cell biology and biochemical research. Its unique attributes position it as a valuable asset in the exploration of cellular membranes, membrane proteins, and intracellular components. Digitonin′s key properties and applications include its ability to solubilize membrane-bound proteins effectively while preserving their native conformation, making it an ideal tool for studying membrane protein interactions and activities.

Moreover, Digitonin selectively permeabilizes the cholesterol-rich plasma membrane, leaving organelle membranes intact. This feature facilitates the extraction and purification of specific organelles, contributing to the study of their functions and roles in cellular processes. The detergent′s capability to permeabilize diverse cell types allows the study of intracellular components and processes by introducing molecules, antibodies, or enzymes into cells. Additionally, Digitonin finds application in immunocytochemistry experiments, enabling the labeling and detection of intracellular proteins and structures.

Application

Digitonin has been used along with permeabilization buffer for cyclic guanosine monophosphate-adenosine monophosphate (cyclic GMP-AMP) stimulation in cells. Digitonin has been used in a study to assess a procedure for isolating mitochondria from ascites tumor cells. It has also been used in a study to investigate its effect on the cytotoxicity of plant secondary metabolites in cancer cells.

Features and Benefits

Highly versatile compound for your cell biology and biochemical research

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service