Skip to Content
Merck
All Photos(1)

Documents

QC1130

Supelco

Demand-WP

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

packaging

ampule of 20 mL

manufacturer/tradename

RTC QCI-026

concentration

100% in H2O

technique(s)

TOC: suitable

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

Product is suitable for measurement of:
  • Biochemical oxygen demand (BOD)
  • Carbonaceous BOD (CBOD)
  • Chemical oxygen demand (COD)
  • Dissolved organic carbon (DOC)
  • Total organic carbon (TOC)

Preparation Note

The solvent for this sample is water. This sample is packaged as a 20 mL concentrate. The sample has been autoclaved for preservation.
PREPARATION INSTRUCTIONS
To prepare solution, partially fill a 1-L volumetric flask with reagent-grade water. Open ampule QC1130-20ML (be careful to avoid cuts on glass) and transfer 10 mL from the ampule into the flask. Fill the volumetric flask to volume with reagent-grade water and mix well. The sample is now ready for immediate analysis.
Analyze the solution to determine BOD or CBOD first. If COD and TOC cannot be determined within two hours after sample preparation, an aliquot should be preserved (pH<2) using 0.2 mL of concentrated sulfuric acid per 100 mL. This aliquot should be stored at 4 degrees Celsius and analyzed as soon as possible within the 28-day maximum holding time.
NOTES: For proper oxygen depletion of BOD and CBOD, the sample should be tested in a dilution series. Use a natural surface water or diluted domestic sewage for seeded dilution water and avoid entrainment of air. For analysis of CBOD, the use of a nitrifying inhibitor is recommended. Be sure to determine the “blank” BOD of your seeded dilution water so that the proper seed correction can be made.

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    5-Day BOD 18 - 230 mg/L
    Carbonaceous BOD (CBOD) 18 - 230 mg/L
    Chemical oxygen demand (COD) 30 - 250 mg/L
    Dissolved organic carbon (DOC) 6 - 100 mg/L
    Total organic carbon (TOC) 6 - 100 mg/L

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service