Skip to Content
Merck
All Photos(1)

Documents

PHR1635

Supelco

Oxcarbazepine

Pharmaceutical Secondary Standard; Certified Reference Material

Synonym(s):

Oxcarbazepine, 10,11-Dihydro-10-oxo-5h-dibenz[b,f]azepine-5-carboxamide, Oxacarbazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12N2O2
CAS Number:
Molecular Weight:
252.27
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to Ph. Eur. Y0001576
traceable to USP 1483152

API family

oxcarbazepine

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1CC2=C(C=CC=C2)N(C(N)=O)C3=CC=CC=C31

InChI

1S/C15H12N2O2/c16-15(19)17-12-7-3-1-5-10(12)9-14(18)11-6-2-4-8-13(11)17/h1-8H,9H2,(H2,16,19)

InChI key

CTRLABGOLIVAIY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Oxcarbazepine is structurally a derivative of carbamazepine and is widely used as an anticonvulsant, antiepileptic and mood stabilizing drug. It is primarily used in the treatment of anxiety, mood disorders, epilepsy and bipolar disorders.

Application

Oxcarbazepine has been used as a pharmaceutical secondary standard for the determination of the analyte in pharmaceutical formulations using spectrophotometric technique and high-performance thin layer chromatography.
These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

Anticonvulsant, antineuralgic. Inhibits veratrine-induced transmitter release.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC1421 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and validation of stability indicating HPTLC method for determination of oxcarbazepine in bulk and pharmaceutical formulation
BhoiteSD, et al.
International Journal of Pharmacy and Pharmaceutical Sciences, 5(3), 127-132 (2013)
Development and validation of stability indicating spectrophotometric methods for determination of oxcarbazepine in pharmaceuticals
Basavaiah K, et al.
J. Sci. Ind. Res., 70, 346-351 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service