Skip to Content
Merck
All Photos(1)

Key Documents

P6430

Sigma-Aldrich

PBFI

~80% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H46N2O16
CAS Number:
Molecular Weight:
882.86
MDL number:
UNSPSC Code:
12171500
Pricing and availability is not currently available.

Assay

~80% (HPLC)

storage temp.

−20°C

SMILES string

COc1cc2cc(oc2cc1N3CCOCCOCCN(CCOCCOCC3)c4cc5oc(cc5cc4OC)-c6ccc(cc6C(O)=O)C(O)=O)-c7ccc(cc7C(O)=O)C(O)=O

Application

Cell impermeable fluorescent probe for K+

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mehdi Rahimi et al.
Luminescence : the journal of biological and chemical luminescence, 34(4), 402-406 (2019-04-30)
Photophysical phenomena associated with carbon nanoparticles in combination with biocompatibility and readily functionalizable properties have attracted significant interest for sensing and imaging applications. A potassium ion optode based on the fluorescence quenching of carbon quantum dots (CQDs) was constructed. The
Ahmed G Abdelhamid et al.
Applied and environmental microbiology, 85(14) (2019-05-19)
Salmonella enterica is increasingly linked to disease outbreaks associated with consumption of low-water-activity (low-aw) foods. Persistence of the pathogen in these foods was attributed to its ability to implement desiccation resistance mechanisms. Published knowledge about methods that disrupt desiccation resistance
Kerstin Boengler et al.
British journal of pharmacology, 174(13), 2060-2073 (2017-04-04)
Dysregulation of gap junction-mediated cell coupling contributes to development of arrhythmias and myocardial damage after ischaemia/reperfusion (I/R). Connexin 43 (Cx43) is present at ventricular gap junctions and also in the mitochondria of cardiomyocytes. The dipeptide (2S, 4R)-1-(2-aminoacetyl)-4-benzamidopyrrolidine-2-carboxylic acid (ZP1609) has
Mikail Temirel et al.
Micromachines, 12(2) (2021-03-07)
Hemp is a sustainable, recyclable, and high-yield annual crop that can be used to produce textiles, plastics, composites, concrete, fibers, biofuels, bionutrients, and paper. The integration of microfluidic paper-based analytical devices (µPADs) with hemp paper can improve the environmental friendliness
Lars F Olsen et al.
Archives of biochemistry and biophysics, 681, 108257-108257 (2020-01-10)
We measured temporal oscillations of intracellular K+ concentration in yeast cells exhibiting glycolytic oscillations using fluorescence spectroscopy and microscopy methods. These oscillations showed the same period as those of glycolytic metabolites (NADH, ATP), indicating a strong coupling between them. We

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service