Skip to Content
Merck
All Photos(1)

Documents

NIST3185

Nitrate anion standard solution

NIST® SRM® 3185

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

packaging

pkg of 5 x 10 mL

manufacturer/tradename

NIST®

format

single component solution

General description

SRM 3185 consists of five sealed borosilicate glass ampoules, each containing 10 mL of solution that has been prepared gravimetrically to contain a known mass fraction of nitrate. The nitrate is dissolved in filtered (0.22 μm) water, which has a minimum resistivity of 18μΩ cm.

SRM 3185_cert

SRM 3185_SDS

Application

Nitrate anion standard solution standard reference material (SRM) is intended for use as a primary calibration standard for the quantitative determination of nitrate using anion ion chromatography (IC).

Features and Benefits

  • Available with a certified mass fraction value of nitrate anion standard solution. Certified values are measured by,
1.Gravimetric preparation using high-purity sodium nitrate. 
2. Anion chromatography calibrated using independently prepared sodium nitrate and potassium nitrate primary standard solutions.
  • Certified values are traceable to the International System of Units (SI) and SI-derived unit of mass fraction. 
  • The NIST certificate is provided with storage, handling, and use instructions.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Nitrate (NO3-)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service