NIST2637A
Carbon monoxide in nitrogen
NIST® SRM® 2637a, nominal 2500 umol/mol
About This Item
Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of cylinder
manufacturer/tradename
NIST®
application(s)
air monitoring
environmental
format
mixture
Looking for similar products? Visit Product Comparison Guide
General description
SRM 2637A_cert
SRM 2637A_SDS
Other Notes
Carbon monoxide (CO)
See certificate for values and more details at nist.gov/SRM.
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Press. Gas Liquefied gas
Storage Class Code
2A - Gases
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service