H0420000
Hexylresorcinol
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
4-Hexylresorcinol, 4-Hexyl-1,3-dihydroxybenzene
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
hexylresorcinol
manufacturer/tradename
EDQM
bp
333-335 °C (lit.)
mp
65-67 °C (lit.)
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
CCCCCCc1ccc(O)cc1O
InChI
1S/C12H18O2/c1-2-3-4-5-6-10-7-8-11(13)9-12(10)14/h7-9,13-14H,2-6H2,1H3
InChI key
WFJIVOKAWHGMBH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Packaging
Other Notes
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
388.4 °F
Flash Point(C)
198 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service