Skip to Content
Merck
All Photos(1)

Key Documents

F8379

Sigma-Aldrich

Fast Red KL Salt

Synonym(s):

2-Carbamoyl-5-methoxybenzenediazonium chloride hemi(zinc chloride) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H8N3O2 · 1/2ZnCl4
CAS Number:
Molecular Weight:
281.77
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

composition

Dye content, ~25%

SMILES string

COc1ccc(C(N)=O)c(c1)[N]#N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service