Skip to Content
Merck
All Photos(1)

Documents

ERMBD518

Bran breakfast cereal (dietary fibre)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material

Agency

ERM®
certified by the BSC

manufacturer/tradename

JRC

application(s)

food and beverages

format

matrix material

storage temp.

−20°C

Analysis Note

For more information please see:
ERMBD518

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Dietary Fibre: Dietary fibre according to AOAC 1990985.29, Dietary fibre according to AOAC 1992 MES-TRIS 991.43, Dietary fibre according to Englyst (by Colorimetry), Dietary fibre according to Englyst (by GC), Dietary fibre according to Uppsala 994.13

Matrix Group: Cereals, Bread, Rice and Maize

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service