Skip to Content
Merck
All Photos(1)

Documents

ERMAD149

Thromboplastin rabbit (prothrombin time)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

ERM®
certified by the BSC

manufacturer/tradename

JRC

application(s)

clinical testing

format

matrix material

shipped in

wet ice

storage temp.

−70°C

Analysis Note

For more information please see:
ERMAD149

Other Notes

Sales restrictions may apply

Legal Information

ERM is a registered trademark of European Commission

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service