Skip to Content
Merck
All Photos(1)

Key Documents

CRM005

Supelco

Trace Metals - Sewage Amended Soil

certified reference material, pkg of 50 g

Synonym(s):

Soil matrix CRM, trace metals

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

certificate is enclosed in each package

packaging

pkg of 50 g

manufacturer/tradename

RTC CRM005-050

technique(s)

AAS: suitable
ICP: suitable

pH

7.59

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
This product was produced and certified from a naturally contaminated matrix and has not been fortified or modified in any way. Analytical data for Certification was obtained using USEPA SW846, 3rd edition methods 3050 (hot block) and 3051(microwave) using nitric acid extraction. Analysis was carried out according to USEPA methods 6010 (ICP-ES), 6020 (ICP-MS) and 7000 (AES).

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

Analyte

Description

    Aluminum
    Arsenic
    Barium
    Beryllium
    Cadmium
    Calcium
    Chromium
    Cobalt
    Copper
    Iron
    Lead
    Magnesium
    Manganese
    Mercury
    Molybdenum
    Nickel
    Potassium
    Selenium
    Silver
    Sodium
    Vanadium
    Zinc
    See All (22)

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service