Skip to Content
Merck
All Photos(1)

Key Documents

BCR549

Acetone-2,4-dinitrophenylhydrazone

BCR®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
238.20
Beilstein:
960878
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.02

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

mp

126-128 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

C\C(C)=N\Nc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C9H10N4O4/c1-6(2)10-11-8-4-3-7(12(14)15)5-9(8)13(16)17/h3-5,11H,1-2H3

InChI key

YGIXYAIGWMAGIB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

For more information please see:
BCR549

Legal Information

BCR is a registered trademark of European Commission

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service