Skip to Content
Merck
All Photos(1)

Key Documents

BCR326

Unalloyed zinc (disc) (trace elements)

BCR®, certified reference material

Synonym(s):

Zinc, unalloyed

Sign Into View Organizational & Contract Pricing

Select a Size

0.7 KG
€347.00

€347.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
0.7 KG
€347.00

About This Item

EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

€347.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material

Agency

BCR®

manufacturer/tradename

JRC

format

matrix material

Analysis Note

For more information please see:
BCR326

Legal Information

BCR is a registered trademark of European Commission

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service