Skip to Content
Merck
All Photos(1)

Documents

90946

Supelco

(1-Cyanocyclohexyl)acetic acid

analytical standard

Synonym(s):

1-Cyanocyclohexaneacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13NO2
CAS Number:
Molecular Weight:
167.21
Beilstein:
11150802
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

form

flakes

shelf life

limited shelf life, expiry date on the label

storage temp.

2-8°C

SMILES string

OC(=O)CC1(CCCCC1)C#N

InChI

1S/C9H13NO2/c10-7-9(6-8(11)12)4-2-1-3-5-9/h1-6H2,(H,11,12)

InChI key

YGFXLCAKCKPSQQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service