Skip to Content
Merck
All Photos(1)

Key Documents

90744

Supelco

Pentyl 4-hydroxybenzoate

analytical standard

Synonym(s):

4-Hydroxybenzoic acid 4-pentyl ester, Amyl p-hydroxybenzoate, NSC 309817, Pentyl paraben

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16O3
CAS Number:
Molecular Weight:
208.25
Beilstein:
2722438
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

format

neat

storage temp.

2-8°C

SMILES string

O=C(OCCCCC)C1=CC=C(O)C=C1

InChI

1S/C12H16O3/c1-2-3-4-9-15-12(14)10-5-7-11(13)8-6-10/h5-8,13H,2-4,9H2,1H3

InChI key

ZNSSPLQZSUWFJT-UHFFFAOYSA-N

General description

Pentyl 4-hydroxybenzoate belongs to the class of parabens, a group of alkyl esters derived from p-hydroxybenzoic acid. They find a variety of applications as preservatives in the cosmetics, pharmaceuticals and food industries.[1]

Application

Pentyl 4-hydroxybenzoate may be used as an analytical reference standard for the quantification of the analyte in cosmetics,[1] water and urine samples[2] using chromatography techniques.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana Victoria Marta-Sanchez et al.
Environmental science and pollution research international, 25(15), 14460-14470 (2018-03-12)
Parabens have been widely used as antimicrobial agents, mainly in food products, pharmaceuticals, and cosmetics. Although they are known as safe preservatives, they also cause some harm to human health, which has been discussed lately. Therefore, the aim of this
Screening of preservatives by HPLC-PDA-ESI/MS: A focus on both allowed and recently forbidden compounds in the new EU cosmetics regulation
Lecce R, et al.
Journal of Pharmaceutical and Biomedical Analysis, 125, 260-269 (2016)
Marta Pastor-Belda et al.
Journal of chromatography. A, 1564, 102-109 (2018-06-17)
This study describes the use of a magnetic multi-walled carbon nanotube composite for the preconcentration of nine parabens (methyl, ethyl, propyl, isopropyl, butyl, isobutyl, pentyl, phenyl and benzylparaben) from water and urine samples after in situ acetylation. The enriched extracts

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service