Skip to Content
Merck
All Photos(2)

Key Documents

901808

Sigma-Aldrich

GreenCat iPr

≥95%

Synonym(s):

(1,3-Bis(2,6-diisopropylphenyl)imidazolidin-2-ylidene)dichloro(2-((1-(methoxy(methyl)amino)-3-methyl-1-oxobutan-2-yl)oxy)benzylidene)ruthenium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H57Cl2N3O3Ru
Molecular Weight:
811.89
UNSPSC Code:
12352101

Assay

≥95%

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

mp

>300 °C

storage temp.

2-8°C

Application

GreenCat is excellent for CM (cross metathesis) of internal, type 1 olefins and is good for CM (cross metathesis) with electron deficient partners.The catalyst is also very good for RCM (ring closing metathesis) with sterically non-demanding substrates leading to small/medium rings. Note: GreenCat is suitable for reaction in "green solvents" without degassing and drying.

Legal Information

Sold in collaboration with Apeiron Synthesis, Inc. Research quantities are 100 g / customer / year. For quantities that exceed the research limit, please contact the supplier at www.apeiron-synthesis.com.
PCT/EP2013/062435

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service