Skip to Content
Merck
All Photos(2)

Documents

89065

Supelco

Stearic acid-13C18

analytical standard

Synonym(s):

Octadecanoic acid-13C18, Stearic acid-ul-13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH3(13CH2)1613CO2H
CAS Number:
Molecular Weight:
302.35
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

isotopic purity

≥99 atom % 13C

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤0.5% water

bp

361 °C (lit.)

mp

68-70 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

functional group

carboxylic acid

mass shift

M+18

shipped in

ambient

storage temp.

room temp

SMILES string

[13CH3][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13CH2][13C](O)=O

InChI

1S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)/i1+1,2+1,3+1,4+1,5+1,6+1,7+1,8+1,9+1,10+1,11+1,12+1,13+1,14+1,15+1,16+1,17+1,18+1

InChI key

QIQXTHQIDYTFRH-HXPQJNGISA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service