Skip to Content
Merck
All Photos(1)

Documents

84380-M

Sigma-Aldrich

Nitric acid

puriss. p.a., ≥65% (T)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HNO3
CAS Number:
Molecular Weight:
63.01
MDL number:
UNSPSC Code:
12352106
eCl@ss:
38060304
PubChem Substance ID:
NACRES:
NA.21

vapor pressure

8 mmHg ( 20 °C)

grade

puriss. p.a.

Assay

≥65% (T)

form

liquid

ign. residue

≤0.0005% (as SO4)

pH

<1.0

bp

120.5 °C (lit.)

density

1.37-1.41 g/mL at 20 °C (lit.)

anion traces

chloride (Cl-): ≤0.5 mg/kg
phosphate (PO43-): ≤0.5 mg/kg
sulfate (SO42-): ≤1 mg/kg

cation traces

Ag: ≤0.01 mg/kg
Al: ≤0.05 mg/kg
As: ≤0.01 mg/kg
Ba: ≤0.01 mg/kg
Bi: ≤0.1 mg/kg
Ca: ≤0.5 mg/kg
Cd: ≤0.01 mg/kg
Co: ≤0.01 mg/kg
Cr: ≤0.02 mg/kg
Cu: ≤0.01 mg/kg
Fe: ≤0.2 mg/kg
Hg: ≤0.01 mg/kg
K: ≤0.05 mg/kg
Li: ≤0.01 mg/kg
Mg: ≤0.1 mg/kg
Mn: ≤0.01 mg/kg
Mo: ≤0.02 mg/kg
Na: ≤0.5 mg/kg
Ni: ≤0.02 mg/kg
Pb: ≤0.01 mg/kg
Sr: ≤0.01 mg/kg
Tl: ≤0.05 mg/kg
Zn: ≤0.05 mg/kg

SMILES string

O[N+]([O-])=O

InChI

1S/HNO3/c2-1(3)4/h(H,2,3,4)

InChI key

GRYLNZFGIOXLOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Nitric acid (HNO3) is a highly corrosive strong mineral acid. In HNO3, nitrogen element exhibits +5 oxidation state. 10%(v/v) nitric acid solution is useful for cleaning laboratory glasswares. Reaction of HNO3 with various original and synthetic mineral dust/mineral oxide surfaces was studied in a low-pressure Knudsen reactor operating at 298K.

Application

Nitric acid (HNO3) may be used in the following studies:
  • Oxidation of multiwalled carbon nanotubes (MWCNTs).
  • Removal of transition metal catalyst from the single-walled carbon nanotubes (SWNTs) during the purification of SWNTs.
  • Trace determinations of Pb and Cd in aqueous medium by flame atomic absorption spectrometry.
  • Preparation of lead dioxide (PbO2).

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Met. Corr. 1 - Ox. Liq. 3 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eduardo Carasek et al.
Talanta, 56(1), 185-191 (2008-10-31)
The need for highly reliable methods for the determination of trace elements has been recognised in analytical chemistry and environmental science. A method for the trace analysis of Pb and Cd in natural waters is described. In a preconcentration step

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service