Skip to Content
Merck
All Photos(1)

Key Documents

83547

Sigma-Aldrich

Regulation valve

suitable for 100 mL and 250 mL pressure tins

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
Pricing and availability is not currently available.

quality

suitable for 100 mL and 250 mL pressure tins

General description

All strained parts are of V4A-steel

Other Notes

Sales restrictions may apply

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service