Skip to Content
Merck
All Photos(1)

Key Documents

80718

Supelco

Tutin solution

20 μg/g in acetonitrile (nominal concentration), certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(1aS,1bR,2S,2′R,5R,6S,6aR,7aR,8R)-Hexahydro-1b,6-dihydroxy-6a-methyl-8-(1-methylethenyl)spiro[2,5-methano-7H-oxireno[3,4]cyclopent[1,2-d]oxepin-7,2′-oxiran]-3(2H)-one solution

Sign Into View Organizational & Contract Pricing

Select a Size

0.5 ML
€233.00

€233.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
0.5 ML
€233.00

About This Item

Empirical Formula (Hill Notation):
C15H18O6
CAS Number:
Molecular Weight:
294.30
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€233.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

concentration

20 μg/g in acetonitrile (nominal concentration)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

single component solution

storage temp.

−20°C

SMILES string

O1[C@@]2([C@@H]3O[C@@H]3[C@@]4([C@]2([C@@H]([C@@H]5OC(=O)[C@H]4[C@@H]5C(=C)C)O)C)O)C1

InChI

1S/C15H18O6/c1-5(2)6-7-12(17)20-8(6)9(16)13(3)14(4-19-14)10-11(21-10)15(7,13)18/h6-11,16,18H,1,4H2,2-3H3/t6-,7+,8+,9+,10+,11-,13-,14+,15-/m0/s1

InChI key

CCAZWUJBLXKBAY-ULZPOIKGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to the SI through a primary reference material from an NMI, e.g. NIST, NRC or NMIJ.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

For the analysis of the phytotoxin tutin in honey.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service