Skip to Content
Merck
All Photos(2)

Key Documents

78251

Supelco

(Z)-Guggulsterone

analytical standard

Synonym(s):

(17Z)-Pregna-4,17(20)-diene-3,16-dione, 4,17(20)-trans-Pregnadiene-3,16-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H28O2
CAS Number:
Molecular Weight:
312.45
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥95.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

[H][C@@]12CCC3=CC(=O)CC[C@]3(C)[C@@]1([H])CC[C@]4(C)\C(=C\C)C(=O)C[C@@]24[H]

InChI

1S/C21H28O2/c1-4-16-19(23)12-18-15-6-5-13-11-14(22)7-9-20(13,2)17(15)8-10-21(16,18)3/h4,11,15,17-18H,5-10,12H2,1-3H3/b16-4+/t15-,17+,18+,20+,21-/m1/s1

InChI key

WDXRGPWQVHZTQJ-OSJVMJFVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

(Z)-Guggulsterone is a natural product that lowers cholesterol due to its function as an antagonist ligand for the bile acid receptor. (Z)-Guggulsterone is a nuclear hormone receptor that regulates the transcription of several genes involved in cholesterol metabolism and plays a role in cholesterol level regulation. (Z)-Guggulsterone is also a selective farnesoid X receptor (FXR) modulator.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liping Yang et al.
Biochimica et biophysica acta, 1844(9), 1684-1693 (2014-06-24)
Farnesoid X receptor (FXR) is a member of the nuclear receptor superfamily of transcription factors that plays a key role in the regulation of bile acids, lipid and glucose metabolisms. The regulative function of FXR is governed by conformational changes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service