Skip to Content
Merck
All Photos(1)

Key Documents

77416

Supelco

Fructone

analytical standard

Synonym(s):

Ethyl 2-methyl-1,3-dioxolane-2-acetate, 2-Methyl-1,3-dioxolane-2-acetic acid ethyl ester, Applinal, Ethyl 2-methyldioxolan-2-yl acetate, Jasmaprunat, NSC 6537

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14O4
CAS Number:
Molecular Weight:
174.19
Beilstein:
130164
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.431-1.435

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

O=C(OCC)CC1(C)OCCO1

InChI

1S/C8H14O4/c1-3-10-7(9)6-8(2)11-4-5-12-8/h3-6H2,1-2H3

InChI key

XWEOGMYZFCHQNT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fructone is an aromatic compound with a strong fruity odor. The olfactory factor is described as pineapple-, strawberry- and apple-like. It is primarily utilized as a fragrance and flavoring material in cosmetics, food and beverages, manufacture of drugs and detergents, and perfume industries.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

199.4 °F

Flash Point(C)

93 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of fructone catalyzed by aluminium sulfate in ionic liquid medium
Lin Q, et al.
Journal of Saudi Chemical Society, 15(2), 101-103 (2011)
Sophia C Poletti et al.
Perception, 46(3-4), 343-351 (2016-10-06)
Background Repeated short-term exposure to odors is known to improve olfaction in patients with acquired olfactory dysfunction. The aim was to find out whether differences in molecular weight of odors used for olfactory training influences olfaction. We hypothesized a greater

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service