Skip to Content
Merck
All Photos(1)

Key Documents

71241

Sigma-Aldrich

Oxalyl chloride

puriss., ≥99.0% (GC)

Synonym(s):

Ethanedioyl dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCOCOCl
CAS Number:
Molecular Weight:
126.93
Beilstein:
1361988
EC Number:
MDL number:
UNSPSC Code:
12352000

vapor density

4.4 (vs air)

vapor pressure

150 mmHg ( 20 °C)

grade

puriss.

Assay

≥99.0% (GC)

reaction suitability

reagent type: oxidant

refractive index

n20/D 1.429 (lit.)

bp

62-65 °C (lit.)

mp

−10-−8 °C (lit.)

density

1.5 g/mL at 20 °C (lit.)

SMILES string

ClC(=O)C(Cl)=O

InChI

1S/C2Cl2O2/c3-1(5)2(4)6

InChI key

CTSLXHKWHWQRSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Synthesis of N-heterocyclic ynones and ynediones, used to activate carboxylic acids
  • Chlorination and halogenation
  • Three-component [3+2] cycloadditions
  • Reactions with organostannanes
  • Synthesis of cyclopentenones
  • Carbonylations, used as a carbonyl synthon

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service