Skip to Content
Merck
All Photos(1)

Key Documents

69867

Sigma-Aldrich

Montmorillonite

KSF, powder

Synonym(s):

K-Catalyst

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352106

form

powder

shelf life

limited shelf life

concentration

≤12% humidity

impurities

8-12% free acid

storage temp.

−20°C

InChI

1S/2Al.4O2Si.H2O.3O/c;;4*1-3-2;;;;/h;;;;;;1H2;;;/q2*+3;;;;;;3*-2

InChI key

GUJOJGAPFQRJSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

surface partially contains H2SO4

Application

catalyst for etherifications/esterifications

Other Notes

Solid Lewis acid catalyst

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Laszlo
Science (New York, N.Y.), 235(4795), 1473-1477 (1987-03-20)
Layer aluminosilicates catalyze reactions in numerous ways. They stabilize high-energy intermediates. They can store energy in their lattice structures and can release it in the form of chemical energy. They can catalyze redox reactions and can serve as photocatalytic devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service