66746
Dichloromethane
suitable for HPLC, ≥99.8% (GC)
Synonym(s):
Methylene chloride
About This Item
6.83 psi ( 20 °C)
Recommended Products
grade
HPLC grade
Quality Level
vapor density
2.9 (vs air)
vapor pressure
24.45 psi ( 55 °C)
6.83 psi ( 20 °C)
Assay
≥99.8% (GC)
form
liquid
autoignition temp.
1223 °F
shelf life
limited shelf life, expiry date on the label
contains
~0.0025% amylene as stabilizer
expl. lim.
22 %
technique(s)
HPLC: suitable
impurities
≤0.001% free acid (as HCl)
≤0.02% water
evapn. residue
≤0.001%
transmittance
232 nm, ≥10%
240 nm, ≥50%
245 nm, ≥80%
260 nm, ≤98%
refractive index
n20/D 1.424 (lit.)
n20/D 1.424
bp
39.8-40 °C (lit.)
mp
−95 °C (lit.)
density
1.325 g/mL at 25 °C (lit.)
solvent strength ε (Al2O3)
0.42 (Lit.)
λ
1 cm path, H2O reference
UV absorption
λ: 232 nm Amax: 1.00
λ: 240 nm Amax: 0.31
λ: 245 nm Amax: 0.10
λ: 260 nm Amax: 0.01
storage temp.
2-8°C
SMILES string
ClCCl
InChI
1S/CH2Cl2/c2-1-3/h1H2
InChI key
YMWUJEATGCHHMB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects
WGK
WGK 2
Flash Point(F)
does not flash
Flash Point(C)
does not flash
Personal Protective Equipment
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH Annex XVII (Restriction List)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service