Skip to Content
Merck
All Photos(1)

Documents

55392

Supelco

1-Hepten-3-ol

analytical standard

Synonym(s):

Butyl vinyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(OH)CH=CH2
CAS Number:
Molecular Weight:
114.19
Beilstein:
1720340
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Quality Level

Assay

≥97.0% (GC)

shelf life

limited shelf life, expiry date on the label

refractive index

n20/D 1.433 (lit.)
n20/D 1.433

bp

153-156 °C (lit.)

density

0.836 g/mL at 20 °C
0.836 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CCCCC(O)C=C

InChI

1S/C7H14O/c1-3-5-6-7(8)4-2/h4,7-8H,2-3,5-6H2,1H3

InChI key

PZKFYTOLVRCMOA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Hepten-3-ol, an allylic alcohol, belongs to the class of volatile organic compounds (VOCs). It is commonly detected as a constituent of Hyacinthus orientalis L. flowers.

Application

1-Hepten-3-ol may be used as an analytical standard for the determination of the analyte in meat products, packaged vegetables, food products, wine, and fruit samples by gas chromatography (GC) based techniques.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

128.8 °F - closed cup

Flash Point(C)

53.8 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Procida et al.
Journal of chromatography. A, 830(1), 175-182 (1999-02-19)
Salami are a typical seasoned sausage of Italy; a number of types are produced, according to local traditional recipes. As industrial production has taken place, a number of problems rise in obtaining products similar to the traditional ones. The use
J L Berdagué et al.
Journal of chromatography. A, 1146(1), 85-92 (2007-02-24)
The aim of this work was to devise a novel fully computerised chromatography-olfactometry system where eight sniffers detect volatile compounds obtained from a single chromatographic separation. The technical options taken to implement this system are described in detail. The new
Quantitative analysis of headspace volatile compounds using comprehensive two-dimensional gas chromatography and their contribution to the aroma of Chardonnay wine
Welke JE, et al.
Food Research International, 59, 85-99 (2014)
Solid phase microextraction/gas chromatography/mass spectrometry integrated with chemometrics for detection of Salmonella typhimurium contamination in a packaged fresh vegetable
Siripatrawan U and Harte BR
Analytica Chimica Acta, 581(1), 63-70 (2007)
K Hosokawa et al.
Plant cell reports, 14(9), 575-579 (1995-06-01)
Essential oils were produced by flowers of Hyacinthus orientalis L. that had been regenerated in vitro. The production of these oils was affected by the concentration of gibberellic acid and sucrose in the medium and by temperature. The highest concentration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service