Skip to Content
Merck
All Photos(1)

Documents

51937

Supelco

N-Acetyl-3,5-dimethyl-4-benzoquinone imine

analytical standard

Synonym(s):

N-(3,5-Dimethyl-4-oxo-2,5-cyclohexadien-1-ylidene)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO2
CAS Number:
Molecular Weight:
177.20
Beilstein:
4982746
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

Assay

≥95.0% (GC)
≥95.0% (elemental analysis)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

storage temp.

−20°C

SMILES string

O=C1C(C)=CC(C=C1C)=NC(C)=O

InChI

1S/C10H11NO2/c1-6-4-9(11-8(3)12)5-7(2)10(6)13/h4-5H,1-3H3

InChI key

VUPORYDINWWWKZ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service