Skip to Content
Merck
All Photos(1)

Key Documents

46340

Sigma-Aldrich

Fisetin

≥99.0% (HPLC)

Synonym(s):

3,3′,4′,7-Tetra­hydroxy­flavone, 5-Deoxyquercetin, Natural Brown 1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O6 · xH2O
CAS Number:
Molecular Weight:
286.24 (anhydrous basis)
Beilstein:
292829
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

≥99.0% (HPLC)

mp

>330 °C (lit.)

SMILES string

O.Oc1ccc2C(=O)C(O)=C(Oc2c1)c3ccc(O)c(O)c3

InChI

1S/C15H10O6.H2O/c16-8-2-3-9-12(6-8)21-15(14(20)13(9)19)7-1-4-10(17)11(18)5-7;/h1-6,16-18,20H;1H2

InChI key

GYHFUROKCOMWNQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sherin Jacob et al.
International journal of developmental neuroscience : the official journal of the International Society for Developmental Neuroscience, 69, 88-96 (2018-07-17)
Methyl mercury is a teratogenic and neurodevelopmental toxicant in the environment. MeHg affects several biological pathways critical for brain development. The present study validated the effect of Fisetin on developmental MeHg exposure induced alterations in mitochondrial apoptotic pathway and Rho
Sherin Jacob et al.
Biological trace element research, 177(2), 297-315 (2016-11-07)
Methyl mercury (MeHg) is a developmental neurotoxin that causes irreversible cognitive damage in offspring of gestationally exposed mothers. Currently, no preventive drugs are established against MeHg developmental neurotoxicity. The neuroprotective effect of gestational administration of a flavanoid against in utero

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service