Skip to Content
Merck
All Photos(1)

Key Documents

44820

Sigma-Aldrich

Eicosane

purum, ≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)18CH3
CAS Number:
Molecular Weight:
282.55
Beilstein:
1700722
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

vapor density

9.8 (vs air)

grade

purum

Assay

≥97.0% (GC)

bp

220 °C/30 mmHg (lit.)

mp

35-37 °C (lit.)
36-38 °C

solubility

hexane: soluble 100 mg/mL, clear

SMILES string

CCCCCCCCCCCCCCCCCCCC

InChI

1S/C20H42/c1-3-5-7-9-11-13-15-17-19-20-18-16-14-12-10-8-6-4-2/h3-20H2,1-2H3

InChI key

CBFCDTFDPHXCNY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Grafting of maleic anhydride to n-eicosane.
Russell KE and Kelusky EC.
Journal of Polymer Science Part A: Polymer Chemistry, 26(8), 2273-2280 (1988)
Wei Wang et al.
Physical chemistry chemical physics : PCCP, 15(34), 14390-14395 (2013-07-25)
Silica microcapsules with hierarchical pore structure are prepared using a one step emulsion templated hydrolysis method. Silica particles of around 100 nm with percolated nanosized pores are self-assembled into micrometer scale spherical shells. The porous structure serves as an ideal
Witold Fortuniak et al.
Colloid and polymer science, 291(3), 725-733 (2013-03-06)
The coemulsification method suitable for the formulation of microcapsules of
B Wenclawiak et al.
Journal of biochemical and biophysical methods, 43(1-3), 197-207 (2000-06-28)
All six insecticide active ingredients in pyrethrum extract were quantified by supercritical fluid chromatography and carbon calibration. Allethrin is a suitable reference compound for carbon calibration and pyrethrins calibrations. Carbon quantification in SFC is also applied to pyrethroids (phenothrin, permethrin
Fernando J Guerrieri et al.
Proceedings. Biological sciences, 276(1666), 2461-2468 (2009-04-15)
Discriminating among individuals and rejecting non-group members is essential for the evolution and stability of animal societies. Ants are good models for studying recognition mechanisms, because they are typically very efficient in discriminating 'friends' (nest-mates) from 'foes' (non-nest-mates). Recognition in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service