Skip to Content
Merck
All Photos(3)

Documents

442828

Sigma-Aldrich

3,6-Dichloro-1,2-benzenedithiol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H2(SH)2
CAS Number:
Molecular Weight:
211.13
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

95%

form

powder, crystals or chunks

impurities

<5% 1,2-benzenedithiol

mp

58-60 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

Sc1c(S)c(Cl)ccc1Cl

InChI

1S/C6H4Cl2S2/c7-3-1-2-4(8)6(10)5(3)9/h1-2,9-10H

InChI key

AJCUDWCLDWDLNY-UHFFFAOYSA-N

Application

3,6-Dichloro-1,2-benzenedithiol has been used as a ligand in thiolate complexes, and a series of homochalcogenide and mixed-chalcogenide ligand complexes.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service